accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHOBr

CAS RN 129722-34-5

IUPAC Name: 7-(4-bromobutoxy)-1,2,3,4-tetrahydroquinolin-2-one
Synonyms: 7-(4-bromobutoxy)-3,4-dihydro-1H-quinolin-2-one
Molecular Weight (g/mol): 298.18
Molecular Formula: C13H16BrNO2
InChi Key: URHLNHVYMNBPEO-UHFFFAOYSA-N
SMILES: BrCCCCOC1=CC2=C(CCC(=O)N2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results