accessibility menu, dialog, popup

UserName

Viewing profile as user:
NO

CAS RN 129747-77-9

IUPAC Name: 3-(1H-pyrrol-1-yl)benzaldehyde
Synonyms: 3-(pyrrol-1-yl)benzaldehyde
Molecular Weight (g/mol): 171.2
Molecular Formula: C11H9NO
InChi Key: PALTUANHIBXQMX-UHFFFAOYSA-N
SMILES: O=CC1=CC(=CC=C1)N1C=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results