accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 129752-86-9

IUPAC Name: chloro[(2,4-dichlorophenyl)methyl]magnesium
Synonyms: chloro[(2,4-dichlorophenyl)methyl]magnesium
Molecular Weight (g/mol): 219.77
Molecular Formula: C7H5Cl3Mg
InChi Key: MJNFFJCHKWQLCN-UHFFFAOYSA-M
SMILES: Cl[Mg]CC1=C(Cl)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results