accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3NH3C(S)OH

CAS RN 129938-20-1

IUPAC Name: hydrogen dimethyl[(1S)-3-(naphthalen-1-yloxy)-1-phenylpropyl]amine chloride
Synonyms: dapoxetine
Molecular Weight (g/mol): 341.88
Molecular Formula: C21H24ClNO
InChi Key: IHWDIQRWYNMKFM-BDQAORGHSA-N
SMILES: [H+].[Cl-].CN(C)[C@@H](CCOC1=C2C=CC=CC2=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Product Line 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)