accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 130-13-2

IUPAC Name: sodium 4-aminonaphthalene-1-sulfonate
Synonyms: sodium 4-aminonaphthalene-1-sulfonate
Molecular Weight (g/mol): 245.23
Molecular Formula: C10H8NNaO3S
InChi Key: JWSRMCCRAJUMLX-UHFFFAOYSA-M
SMILES: [Na+].NC1=C2C=CC=CC2=C(C=C1)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results