accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOSOO

CAS RN 130-14-3

IUPAC Name: sodium naphthalene-1-sulfonate
Synonyms: sodium 1-naphthalenesulfonate
Molecular Weight (g/mol): 230.21
Molecular Formula: C10H7NaO3S
InChi Key: HIEHAIZHJZLEPQ-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)