CAS RN 130-15-4
1,4-Naphthoquinone 98.0+%, TCI America™
CAS: 130-15-4 Molecular Formula: C10H6O2 Molecular Weight (g/mol): 158.156 MDL Number: MFCD00001676 InChI Key: FRASJONUBLZVQX-UHFFFAOYSA-N Synonym: 1,4-naphthoquinone,1,4-naphthalenedione,p-naphthoquinone,naphthoquinone,alpha-naphthoquinone,1,4-naphthylquinone,usaf cy-10,1,4-dihydronaphthalene-1,4-dione,1,4-dihydro-1,4-diketonaphthalene,1,4-naftochinon PubChem CID: 8530 ChEBI: CHEBI:27418 IUPAC Name: naphthalene-1,4-dione SMILES: C1=CC=C2C(=O)C=CC(=O)C2=C1
1,4-Naphthoquinone, 99%, contains up to 6% water
CAS: 130-15-4 Molecular Formula: C10H6O2 Molecular Weight (g/mol): 158.16 MDL Number: MFCD00001676 InChI Key: FRASJONUBLZVQX-UHFFFAOYSA-N Synonym: 1,4-naphthoquinone,1,4-naphthalenedione,p-naphthoquinone,naphthoquinone,alpha-naphthoquinone,1,4-naphthylquinone,usaf cy-10,1,4-dihydronaphthalene-1,4-dione,1,4-dihydro-1,4-diketonaphthalene,1,4-naftochinon PubChem CID: 8530 ChEBI: CHEBI:27418 IUPAC Name: naphthalene-1,4-dione SMILES: C1=CC=C2C(=O)C=CC(=O)C2=C1
1,4-Naphthoquinone, 97+% (dry wt.), cont. up to 5% water
CAS: 130-15-4 Molecular Formula: C10H6O2 Molecular Weight (g/mol): 158.156 MDL Number: MFCD00001676 InChI Key: FRASJONUBLZVQX-UHFFFAOYSA-N Synonym: 1,4-naphthoquinone,1,4-naphthalenedione,p-naphthoquinone,naphthoquinone,alpha-naphthoquinone,1,4-naphthylquinone,usaf cy-10,1,4-dihydronaphthalene-1,4-dione,1,4-dihydro-1,4-diketonaphthalene,1,4-naftochinon PubChem CID: 8530 ChEBI: CHEBI:27418 IUPAC Name: naphthalene-1,4-dione SMILES: C1=CC=C2C(=O)C=CC(=O)C2=C1
Sigma Aldrich 1,4-Naphthoquinone
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 1,4-Naphthoquinone
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
1,4-Napthoquinone, SPEX CertiPrep™
CAS: 130-15-4 Molecular Formula: C10H6O2 Molecular Weight (g/mol): 158.156 InChI Key: FRASJONUBLZVQX-UHFFFAOYSA-N PubChem CID: 8530 ChEBI: CHEBI:27418 IUPAC Name: naphthalene-1,4-dione SMILES: C1=CC=C2C(=O)C=CC(=O)C2=C1