CAS RN 130-16-5
5-Chloro-8-hydroxyquinoline, 98%
CAS: 130-16-5 Molecular Formula: C9H6ClNO Molecular Weight (g/mol): 179.61 MDL Number: MFCD00006788 InChI Key: CTQMJYWDVABFRZ-UHFFFAOYSA-N Synonym: 5-chloro-8-hydroxyquinoline,cloxiquine,cloxyquin,5-chloro-8-quinolinol,dermofongin,5-chlorooxine,8-quinolinol, 5-chloro,chlorisept,dermofungin,5-chloro-8-oxyquinoline PubChem CID: 2817 IUPAC Name: 5-chloroquinolin-8-ol SMILES: C1=CC2=C(C=CC(=C2N=C1)O)Cl
5-Chloro-8-hydroxyquinoline, 95%
CAS: 130-16-5 Molecular Formula: C9H6ClNO Molecular Weight (g/mol): 179.603 MDL Number: MFCD00006788 InChI Key: CTQMJYWDVABFRZ-UHFFFAOYSA-N Synonym: 5-chloro-8-hydroxyquinoline,cloxiquine,cloxyquin,5-chloro-8-quinolinol,dermofongin,5-chlorooxine,8-quinolinol, 5-chloro,chlorisept,dermofungin,5-chloro-8-oxyquinoline PubChem CID: 2817 IUPAC Name: 5-chloroquinolin-8-ol SMILES: C1=CC2=C(C=CC(=C2N=C1)O)Cl
5-Chloro-8-hydroxyquinoline 98.0+%, TCI America™
CAS: 130-16-5 Molecular Formula: C9H6ClNO Molecular Weight (g/mol): 179.603 MDL Number: MFCD00006788 InChI Key: CTQMJYWDVABFRZ-UHFFFAOYSA-N Synonym: 5-chloro-8-hydroxyquinoline,cloxiquine,cloxyquin,5-chloro-8-quinolinol,dermofongin,5-chlorooxine,8-quinolinol, 5-chloro,chlorisept,dermofungin,5-chloro-8-oxyquinoline PubChem CID: 2817 IUPAC Name: 5-chloroquinolin-8-ol SMILES: C1=CC2=C(C=CC(=C2N=C1)O)Cl
Sigma Aldrich 5-Chloro-8-quinolinol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc 5-Chloro-8-Hydroxyquinoline, Approx. 96%, MP Biomedicals™
CAS: 130-16-5 Molecular Formula: C9H6ClNO Molecular Weight (g/mol): 179.603 InChI Key: CTQMJYWDVABFRZ-UHFFFAOYSA-N Synonym: 5-chloro-8-hydroxyquinoline,cloxiquine,cloxyquin,5-chloro-8-quinolinol,dermofongin,5-chlorooxine,8-quinolinol, 5-chloro,chlorisept,dermofungin,5-chloro-8-oxyquinoline PubChem CID: 2817 IUPAC Name: 5-chloroquinolin-8-ol SMILES: C1=CC2=C(C=CC(=C2N=C1)O)Cl
MP Biomedicals, Inc 5-Chloro-8-hydroxyquinoline, MP Biomedicals
CAS: 130-16-5 Molecular Formula: C9H6ClNO Molecular Weight (g/mol): 179.603 InChI Key: CTQMJYWDVABFRZ-UHFFFAOYSA-N Synonym: 5-chloro-8-hydroxyquinoline,cloxiquine,cloxyquin,5-chloro-8-quinolinol,dermofongin,5-chlorooxine,8-quinolinol, 5-chloro,chlorisept,dermofungin,5-chloro-8-oxyquinoline PubChem CID: 2817 IUPAC Name: 5-chloroquinolin-8-ol SMILES: C1=CC2=C(C=CC(=C2N=C1)O)Cl