CAS RN 130-95-0
Quinine, anhydrous, 99% (total base), may cont. up to 5% dihydroquinine
CAS: 130-95-0 Molecular Formula: C20H24N2O2 Molecular Weight (g/mol): 324.42 MDL Number: MFCD00198096 InChI Key: LOUPRKONTZGTKE-ZCXAXGJJNA-N Synonym: 1r-6-methoxyquinolin-4-yl 1s,4s,5r-5-vinylquinuclidin-2-yl methanol,quinine anhydrous,+-2,3-o-isopropylidene-l-threitol PubChem CID: 129316724 SMILES: [H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
Quinine, 98+%, anhydrous
CAS: 130-95-0 Molecular Formula: C20H24N2O2 Molecular Weight (g/mol): 324.42 MDL Number: MFCD00198096 InChI Key: LOUPRKONTZGTKE-ZCXAXGJJNA-N Synonym: 1r-6-methoxyquinolin-4-yl 1s,4s,5r-5-vinylquinuclidin-2-yl methanol,quinine anhydrous,+-2,3-o-isopropylidene-l-threitol PubChem CID: 129316724 IUPAC Name: (5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl)-[6-(trideuteriomethoxy)quinolin-4-yl]methanol SMILES: [H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
phyproof™ (-)-Quinine, ≥90% (HPLC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Quinine, 95%, MP Biomedicals™
CAS: 130-95-0 Molecular Formula: C20H24N2O2 Molecular Weight (g/mol): 324.42 MDL Number: MFCD00198096 InChI Key: LOUPRKONTZGTKE-ZCXAXGJJNA-N Synonym: quinine,chinin,chinine,--quinine,qualaquin,6'-methoxycinchonidine,8s,9r-quinine,chininum,quinine sulfate,quinina PubChem CID: 129316724 IUPAC Name: (R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol SMILES: [H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
Quinine 98.0+%, TCI America™
CAS: 130-95-0 Molecular Formula: C20H24N2O2 Molecular Weight (g/mol): 324.42 MDL Number: MFCD00198096 InChI Key: LOUPRKONTZGTKE-ZCXAXGJJNA-N Synonym: 1r-6-methoxyquinolin-4-yl 1s,4s,5r-5-vinylquinuclidin-2-yl methanol,quinine anhydrous,+-2,3-o-isopropylidene-l-threitol PubChem CID: 129316724 IUPAC Name: (R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol SMILES: [H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
Sigma Aldrich Quinine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.