CAS RN 1300-94-3
IUPAC Name:
5-methyl-2-pentylphenol
Synonyms:
amylmetacresol amylmetacresolum amilmetacresol
Molecular Weight (g/mol):
178.28
Molecular Formula:
C12H18O
InChi Key:
CKGWFZQGEQJZIL-UHFFFAOYSA-N
SMILES:
CCCCCC1=CC=C(C)C=C1O
1
–
1
of
1
results
1
2-Amyl-5-methylphenol 98.0+%, TCI America™
CAS: 1300-94-3 Molecular Formula: C12H18O Molecular Weight (g/mol): 178.275 MDL Number: MFCD00127710 InChI Key: CKGWFZQGEQJZIL-UHFFFAOYSA-N Synonym: 5-Methyl-2-pentylphenol PubChem CID: 14759 ChEBI: CHEBI:48213 IUPAC Name: 5-methyl-2-pentylphenol SMILES: CCCCCC1=C(C=C(C=C1)C)O