accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 130004-33-0

IUPAC Name: 1-methyl-4-(propan-2-yl)benzene; [2'-(diphenylphosphanyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphane; dichlororuthenium
Synonyms: binap; cymene; dichlororuthenium
Molecular Weight (g/mol): 928.88
Molecular Formula: C54H46Cl2P2Ru
InChi Key: WNHLGYRPKARUHY-UHFFFAOYSA-L
SMILES: Cl[Ru]Cl.CC(C)C1=CC=C(C)C=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results