accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 130047-19-7

IUPAC Name: 4-fluoro-2-(trifluoromethyl)phenol
Synonyms: 4-fluoro-2-(trifluoromethyl)phenol
Molecular Weight (g/mol): 180.1
Molecular Formula: C7H4F4O
InChi Key: RNOVGJWJVRESAA-UHFFFAOYSA-N
SMILES: OC1=C(C=C(F)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results