accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13007-92-6

IUPAC Name: hexakis(methanidylidyneoxidanium) chromium
Synonyms: chromium hexacarbonyl
Molecular Weight (g/mol): 220.06
Molecular Formula: C6CrO6
InChi Key: KOTQLLUQLXWWDK-UHFFFAOYSA-N
SMILES: [Cr].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)