accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1301-96-8

IUPAC Name: oxosilver
Synonyms: Silver(I,III) oxide
Molecular Weight (g/mol): 123.87
Molecular Formula: AgO
InChi Key: OTCVAHKKMMUFAY-UHFFFAOYSA-N
SMILES: O=[Ag]
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (6)
  • (2)
  • (2)
  • (3)
  • (3)
  • (8)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)