accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13010-19-0

IUPAC Name: 1-chloro-3-isocyanatopropane
Synonyms: 1-chloro-3-isocyanatopropane
Molecular Weight (g/mol): 119.55
Molecular Formula: C4H6ClNO
InChi Key: RQAVSDINDRNIKL-UHFFFAOYSA-N
SMILES: ClCCCN=C=O
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)