CAS RN 13014-24-9
IUPAC Name:
1,2-dichloro-4-(trichloromethyl)benzene
Synonyms:
1,2-dichloro-4-(trichloromethyl)benzene
Molecular Weight (g/mol):
264.35
Molecular Formula:
C7H3Cl5
InChi Key:
ATYLRBXENHNROH-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1Cl)C(Cl)(Cl)Cl
1
–
1
of
1
results
1
3,4-Dichlorobenzotrichloride 98.0+%, TCI America™
CAS: 13014-24-9 Molecular Formula: C7H3Cl5 Molecular Weight (g/mol): 264.35 MDL Number: MFCD00018820 InChI Key: ATYLRBXENHNROH-UHFFFAOYSA-N Synonym: alpha,alpha,alpha,3,4-Pentachlorotoluene PubChem CID: 25608 IUPAC Name: 1,2-dichloro-4-(trichloromethyl)benzene SMILES: ClC1=CC=C(C=C1Cl)C(Cl)(Cl)Cl