accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 130161-46-5

IUPAC Name: 2-phenylpyrimidine-5-carbaldehyde
Synonyms: 2-phenylpyrimidine-5-carbaldehyde
Molecular Weight (g/mol): 184.2
Molecular Formula: C11H8N2O
InChi Key: AUTGLFSBJLERMV-UHFFFAOYSA-N
SMILES: O=CC1=CN=C(N=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results