accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1301613-77-3

IUPAC Name: 3,6-dibromopyrazine-2-carboxamide
Synonyms: 3,6-dibromopyrazine-2-carboxamide
Molecular Weight (g/mol): 280.91
Molecular Formula: C5H3Br2N3O
InChi Key: TWCACYWIPUIXAR-UHFFFAOYSA-N
SMILES: NC(=O)C1=NC(Br)=CN=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results