accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHHNOOCH3H3CCH3OCH3OOHOOH(E/Z)

CAS RN 1301706-76-2

IUPAC Name: but-2-enedioic acid; tert-butyl N-[(4,4-diethoxypiperidin-2-yl)methyl]carbamate
Synonyms: butenedioic acid; tert-butyl N-[(4,4-diethoxypiperidin-2-yl)methyl]carbamate
Molecular Weight (g/mol): 418.49
Molecular Formula: C19H34N2O8
InChi Key: HCNKZGJFFYKTGU-UHFFFAOYNA-N
SMILES: OC(=O)C=CC(O)=O.CCOC1(CCNC(CNC(=O)OC(C)(C)C)C1)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results