accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3CH3

CAS RN 13019-20-0

IUPAC Name: 2-methylheptan-3-one
Synonyms: 3-heptanone, 2-methyl-
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: XYYMFUCZDNNGFS-UHFFFAOYSA-N
SMILES: CCCCC(=O)C(C)C
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)