accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOAlO

CAS RN 1302-42-7

IUPAC Name: sodium dioxoalumanuide
Synonyms: sodium aluminate
Molecular Weight (g/mol): 81.97
Molecular Formula: AlNaO2
InChi Key: IYJYQHRNMMNLRH-UHFFFAOYSA-N
SMILES: [Na+].O=[Al-]=O
Grade 
  • (4)
  • (2)
Physical Form 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)