accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOH3COH3C(E/Z)(E/Z)(E/Z)

CAS RN 13023-13-7

IUPAC Name: tris(octadec-9-en-1-yl) phosphite
Synonyms: tris(octadec-9-en-1-yl) phosphite
Molecular Weight (g/mol): 833.41
Molecular Formula: C54H105O3P
InChi Key: QULKPMBZSVVAJM-UHFFFAOYSA-N
SMILES: CCCCCCCCC=CCCCCCCCCOP(OCCCCCCCCC=CCCCCCCCC)OCCCCCCCCC=CCCCCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results