accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13028-69-8

IUPAC Name: butyltriethylazanium
Synonyms: butyltriethylazanium
Molecular Weight (g/mol): 158.31
Molecular Formula: C10H24N
InChi Key: MPDDDPYHTMZBMG-UHFFFAOYSA-N
SMILES: CCCC[N+](CC)(CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results