accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrClNH2

CAS RN 130284-53-6

IUPAC Name: 5-bromo-6-chloropyridin-3-amine
Synonyms: 5-bromo-6-chloropyridin-3-amine
Molecular Weight (g/mol): 207.46
Molecular Formula: C5H4BrClN2
InChi Key: ISKBXMMELYRESC-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=C(Cl)N=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results