accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13029-09-9

IUPAC Name: 2,2'-dibromo-1,1'-biphenyl
Synonyms: 2,2'-dibromo-1,1'-biphenyl
Molecular Weight (g/mol): 312
Molecular Formula: C12H8Br2
InChi Key: DRKHIWKXLZCAKP-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1C1=CC=CC=C1Br
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (4)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (4)
  • (3)
  • (1)
  • (3)
  • (3)