accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 130290-79-8

IUPAC Name: (oxan-4-yl)methanaminium
Synonyms: oxan-4-ylmethanaminium
Molecular Weight (g/mol): 116.18
Molecular Formula: C6H14NO
InChi Key: IPBPLHNLRKRLPJ-UHFFFAOYSA-O
SMILES: [NH3+]CC1CCOCC1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (3)
  • (5)
  • (2)
  • (6)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)