accessibility menu, dialog, popup

UserName

Viewing profile as user:
RhSbFFFFFF

CAS RN 130296-28-5

IUPAC Name: λ¹-rhodium(1+) bis((1Z,5Z)-cycloocta-1,5-diene) hexafluorostibanuide
Synonyms: λ¹-rhodium(1+) bis(1,5-cyclooctadiene, (Z,Z)-) hexafluoroantimonate ion
Molecular Weight (g/mol): 555.02
Molecular Formula: C16H24F6RhSb
InChi Key: BROZUPPJWNODCU-SXGOCSEPSA-H
SMILES: [Rh+].F[Sb-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results