accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1303-00-0

IUPAC Name: gallanylidynearsane
Synonyms: gallium arsenide
Molecular Weight (g/mol): 144.65
Molecular Formula: AsGa
InChi Key: JBRZTFJDHDCESZ-UHFFFAOYSA-N
SMILES: [Ga]#[As]
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)