accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1303-36-2

IUPAC Name: [(selanylidenearsanyl)selanyl]arsaneselone
Synonyms: arsenic selenide arsenic triselenide
Molecular Weight (g/mol): 386.76
Molecular Formula: As2Se3
InChi Key: WBFMCDAQUDITAS-UHFFFAOYSA-N
SMILES: [Se]=[As][Se][As]=[Se]
Percent Purity 
  • (4)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results