accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13031-60-2

IUPAC Name: 4-methoxy-4-oxobutan-1-aminium chloride
Synonyms: 4-methoxy-4-oxobutan-1-aminium chloride
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: WPGPRLVPWACBHW-UHFFFAOYSA-N
SMILES: [Cl-].COC(=O)CCC[NH3+]
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)