accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOO

CAS RN 130312-02-6

IUPAC Name: benzyl 3-oxopyrrolidine-1-carboxylate
Synonyms: benzyl 3-oxopyrrolidine-1-carboxylate
Molecular Weight (g/mol): 219.24
Molecular Formula: C12H13NO3
InChi Key: LMHWEUQNJRXMCD-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC=CC=C1)N1CCC(=O)C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)