accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13036-02-7

IUPAC Name: 1,3-dimethyl 5-hydroxybenzene-1,3-dicarboxylate
Synonyms: 1,3-dimethyl 5-hydroxybenzene-1,3-dicarboxylate
Molecular Weight (g/mol): 210.19
Molecular Formula: C10H10O5
InChi Key: DOSDTCPDBPRFHQ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC(O)=C1)C(=O)OC
Molecular Weight (g/mol) 
  • (1)
  • (7)
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (7)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)