accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2COH

CAS RN 13038-21-6

IUPAC Name: non-8-en-1-ol
Synonyms: non-8-en-1-ol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: FKGFCVJJLGSFSB-UHFFFAOYSA-N
SMILES: OCCCCCCCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results