accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2

CAS RN 13038-22-7

IUPAC Name: non-8-en-1-yl acetate
Synonyms: non-8-en-1-yl acetate
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: HUXQMCFHKGATCP-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCCCCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results