accessibility menu, dialog, popup

UserName

Viewing profile as user:
BeO

CAS RN 1304-56-9

IUPAC Name: oxoberyllium
Synonyms: beryllium oxide
Molecular Weight (g/mol): 25.01
Molecular Formula: BeO
InChi Key: LTPBRCUWZOMYOC-UHFFFAOYSA-N
SMILES: [Be]=O
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)