accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1304-82-1

IUPAC Name: dibismuth(3+) tritellanediide
Synonyms: bismuth telluride
Molecular Weight (g/mol): 800.76
Molecular Formula: Bi2Te3
InChi Key: AZFMNKUWQAGOBM-UHFFFAOYSA-N
SMILES: [Te--].[Te--].[Te--].[Bi+3].[Bi+3]
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)