accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOBiONOOONOO

CAS RN 1304-85-4

IUPAC Name: bis(nitrooxy)bismuthanyl nitrate
Synonyms: bismuth subnitrate
Molecular Weight (g/mol): 394.99
Molecular Formula: BiN3O9
InChi Key: PPNKDDZCLDMRHS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)O[Bi](O[N+]([O-])=O)O[N+]([O-])=O
Grade 
  • (2)
  • (7)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (8)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)