accessibility menu, dialog, popup

UserName

Viewing profile as user:
Br(E/Z)

CAS RN 13041-70-8

IUPAC Name: 1-bromo-4-(2-phenylethenyl)benzene
Synonyms: 1-bromo-4-(2-phenylethenyl)benzene
Molecular Weight (g/mol): 259.15
Molecular Formula: C14H11Br
InChi Key: ZZMMKLVIBZWGPK-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=CC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results