accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13048-34-5

IUPAC Name: 10-(prop-2-enoyloxy)decyl prop-2-enoate
Synonyms: 10-(prop-2-enoyloxy)decyl prop-2-enoate
Molecular Weight (g/mol): 282.38
Molecular Formula: C16H26O4
InChi Key: RHNJVKIVSXGYBD-UHFFFAOYSA-N
SMILES: C=CC(=O)OCCCCCCCCCCOC(=O)C=C
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results