accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 130612-84-9

SNF

CAS RN 130612-84-9

IUPAC Name: 2-fluoro-2-(phenylsulfanyl)acetonitrile
Synonyms: 2-fluoro-2-(phenylsulfanyl)acetonitrile
Molecular Weight (g/mol): 167.2
Molecular Formula: C8H6FNS
InChi Key: VAMSBCYZRBSCPJ-UHFFFAOYNA-N
SMILES: FC(SC1=CC=CC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results