accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13064-83-0

IUPAC Name: 4-methylcyclohexane-1-carboxylic acid
Synonyms: 4-methylcyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: QTDXSEZXAPHVBI-UHFFFAOYSA-N
SMILES: CC1CCC(CC1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results