CAS RN 13066-01-8
CAS RN 13066-01-8
IUPAC Name:
4-(2-methoxyphenoxy)aniline
Synonyms:
4-(2-methoxyphenoxy)aniline
Molecular Weight (g/mol):
215.25
Molecular Formula:
C13H13NO2
InChi Key:
XFOFRBMGVDBINH-UHFFFAOYSA-N
SMILES:
COC1=CC=CC=C1OC1=CC=C(N)C=C1
1
–
1
of
1
results
1
Sigma Aldrich 4-(2-Methoxyphenoxy)aniline
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.