accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSiFFFFFFFFFFFFFOCH3OH3C

CAS RN 130676-81-2

IUPAC Name: triethoxy[5,5,6,6,7,7,7-heptafluoro-4,4-bis(trifluoromethyl)heptyl]silane
Synonyms: triethoxy[5,5,6,6,7,7,7-heptafluoro-4,4-bis(trifluoromethyl)heptyl]silane
Molecular Weight (g/mol): 524.39
Molecular Formula: C15H21F13O3Si
InChi Key: JVAFDQZZUMUFSM-UHFFFAOYSA-N
SMILES: CCO[Si](CCCC(C(F)(F)F)(C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)F)(OCC)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results