accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaClOsClClClClCl

CAS RN 1307-81-9

IUPAC Name: disodium hexachloroosmiumdiuide
Synonyms: disodium hexachloroosmiumdiuide
Molecular Weight (g/mol): 448.91
Molecular Formula: Cl6Na2Os
InChi Key: PVTVRPSPEDISMV-UHFFFAOYSA-H
SMILES: [Na+].[Na+].Cl[Os--](Cl)(Cl)(Cl)(Cl)Cl
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results