accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1307-99-9

IUPAC Name: selanylidenecobalt
Synonyms: cobalt(II) selenide
Molecular Weight (g/mol): 137.9
Molecular Formula: CoSe
InChi Key: QVYIMIJFGKEJDW-UHFFFAOYSA-N
SMILES: [Co]=[Se]
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)