accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOH3C

CAS RN 13078-21-2

IUPAC Name: ethyl 2-(2-methoxyphenoxy)acetate
Synonyms: ethyl 2-(2-methoxyphenoxy)acetate
Molecular Weight (g/mol): 210.23
Molecular Formula: C11H14O4
InChi Key: BZCGGCRVJFWCIW-UHFFFAOYSA-N
SMILES: CCOC(=O)COC1=CC=CC=C1OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results