accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1308-38-9

IUPAC Name: dichromium(3+) trioxidandiide
Synonyms: dichromium(3+) trioxidandiide
Molecular Weight (g/mol): 151.99
Molecular Formula: Cr2O3
InChi Key: UOUJSJZBMCDAEU-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Cr+3].[Cr+3]
Quantity 
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (6)
Physical Form 
  • (2)
  • (2)
  • (10)
  • (5)
  • (1)
Grade 
  • (4)
  • (5)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)
  • (13)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (6)
  • (2)
  • (2)
  • (10)
  • (5)
  • (1)
  • (4)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)