CAS RN 13080-85-8
IUPAC Name:
4-{[4'-(4-aminophenoxy)-[1,1'-biphenyl]-4-yl]oxy}aniline
Synonyms:
4-{[4'-(4-aminophenoxy)-[1,1'-biphenyl]-4-yl]oxy}aniline
Molecular Weight (g/mol):
368.44
Molecular Formula:
C24H20N2O2
InChi Key:
HYDATEKARGDBKU-UHFFFAOYSA-N
SMILES:
NC1=CC=C(OC2=CC=C(C=C2)C2=CC=C(OC3=CC=C(N)C=C3)C=C2)C=C1
1
–
1
of
1
results
1
4,4'-Bis(4-aminophenoxy)biphenyl 98.0+%, TCI America™
CAS: 13080-85-8 Molecular Formula: C24H20N2O2 Molecular Weight (g/mol): 368.44 MDL Number: MFCD00039151 InChI Key: HYDATEKARGDBKU-UHFFFAOYSA-N PubChem CID: 632695 IUPAC Name: 4-{[4'-(4-aminophenoxy)-[1,1'-biphenyl]-4-yl]oxy}aniline SMILES: NC1=CC=C(OC2=CC=C(C=C2)C2=CC=C(OC3=CC=C(N)C=C3)C=C2)C=C1