CAS RN 1308074-49-8
IUPAC Name:
2-bromo-N,N-bis(2-methylpropyl)benzene-1-sulfonamide
Synonyms:
2-bromo-N,N-bis(2-methylpropyl)benzenesulfonamide
Molecular Weight (g/mol):
348.3
Molecular Formula:
C14H22BrNO2S
InChi Key:
QYAKCDYMLXIWKK-UHFFFAOYSA-N
SMILES:
CC(C)CN(CC(C)C)S(=O)(=O)C1=CC=CC=C1Br
1
–
1
of
1
results
1
2-Bromo-N,N-diisobutylbenzenesulfonamide, 97%, Thermo Scientific™
CAS: 1308074-49-8 Molecular Formula: C14H22BrNO2S Molecular Weight (g/mol): 348.299 MDL Number: MFCD13225051 InChI Key: QYAKCDYMLXIWKK-UHFFFAOYSA-N Synonym: 2-bromo-n,n-bis 2-methylpropyl benzenesulfonamide,2-bromo-n,n-diisobutylbenzenesulfonamide PubChem CID: 60707653 IUPAC Name: 2-bromo-N,N-bis(2-methylpropyl)benzenesulfonamide SMILES: CC(C)CN(CC(C)C)S(=O)(=O)C1=CC=CC=C1Br