accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1309-36-0

IUPAC Name: disulfanylideneiron
Synonyms: disulfanylideneiron
Molecular Weight (g/mol): 119.97
Molecular Formula: FeS2
InChi Key: NFMAZVUSKIJEIH-UHFFFAOYSA-N
SMILES: S=[Fe]=S
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results